siRNA Details
| Gene Symbol: KCNC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039574.2 | NP_001034663.1 | 2 | 1543 | ||
| NM_004978.4 | NP_004969.2 | 2 | 1543 | |||
| NR_036437.1 | 2 | 1543 | ||||
| XM_006710625.3 | XP_006710688.1 | 2 | 1387 | |||
| XM_006710627.3 | XP_006710690.1 | 2 | 1387 | |||
| XM_011541401.2 | XP_011539703.1 | 2 | 1387 | |||
| GenBank mRNA | AK310193.1 | - | 2 | 968 | ||
| BC101769.1 | - | 2 | 814 | 8069275 | ||
| BX537638.1 | - | 1 | 3331 | |||
| M64676.1 | - | 2 | 913 | |||

KCNC4
Genomic Map