siRNA Details
| Gene Symbol: APPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251904.1 | NP_001238833.1 | 21 | 2141 | ||
| NM_001251905.1 | NP_001238834.1 | 21 | 2166 | |||
| NM_018171.3 | NP_060641.2 | 21 | 2123 | |||
| XM_006719472.1 | XP_006719535.1 | 20 | 1919 | |||
| XM_011538530.2 | XP_011536832.1 | 21 | 3593 | |||
| XM_011538531.2 | XP_011536833.1 | 21 | 3256 | |||
| XM_011538532.2 | XP_011536834.1 | 21 | 4582 | |||
| XM_017019551.1 | XP_016875040.1 | 21 | 2528 | |||
| XM_017019552.1 | XP_016875041.1 | 21 | 3238 | |||
| XM_017019553.1 | XP_016875042.1 | 22 | 4836 | |||
| XM_017019554.1 | XP_016875043.1 | 20 | 1901 | |||

APPL2
Genomic Map