siRNA Details
| Gene Symbol: FAM86C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099653.1 | NP_001093123.1 | 4, 5 | 418 | ||
| NM_018172.2 | NP_060642.2 | 4, 5 | 439 | |||
| NM_152563.2 | NP_689776.1 | 3, 4 | 337 | |||
| GenBank mRNA | AK001523.1 | - | 4, 5 | 413 | ||
| AK130709.1 | - | 3, 4 | 334 | |||
| AK311428.1 | - | 4, 5 | 395 | |||
| AK311518.1 | - | 4, 5 | 418 | |||
| BC032519.1 | - | 3, 4 | 337 | 5494088 | ||
| BQ921333.1 | - | 4, 5 | 388 | |||

FAM86C1
Genomic Map