siRNA Details
| Gene Symbol: PLEKHG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144856.1 | NP_001138328.1 | 7 | 851 | ||
| NM_001144857.1 | NP_001138329.1 | 6 | 1019 | |||
| NM_018173.3 | NP_060643.2 | 7 | 769 | |||
| XM_005253704.4 | XP_005253761.1 | 7 | 866 | |||
| XM_006718985.3 | XP_006719048.1 | 7 | 866 | |||
| XM_011520967.2 | XP_011519269.1 | 7 | 741 | |||
| XM_017019555.1 | XP_016875044.1 | 7 | 866 | |||
| GenBank mRNA | AK001527.1 | - | 7 | 770 | ||
| AK023741.1 | - | 7 | 851 | |||
| AK095373.1 | - | 6 | 1019 | |||
| BC104751.1 | - | 7 | 705 | 40035012 | ||
| BC105961.1 | - | 7 | 705 | 40035014 | ||
| BC109095.2 | - | 7 | 705 | 40034695 | ||

PLEKHG6
Genomic Map