siRNA Details
| Gene Symbol: RCBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018191.3 | NP_060661.3 | 4 | 402 | ||
| XM_005266441.2 | XP_005266498.1 | 3 | 322 | |||
| XM_011535133.1 | XP_011533435.1 | 4 | 428 | |||
| XM_011535134.1 | XP_011533436.1 | 4 | 328 | |||
| XM_011535135.2 | XP_011533437.1 | 3 | 237 | |||
| XM_011535136.2 | XP_011533438.1 | 4 | 406 | |||
| GenBank mRNA | AF334406.1 | - | 4 | 376 | ||
| AJ319660.1 | - | 4 | 343 | |||
| AK096654.1 | - | 4 | 402 | |||
| AL833821.1 | - | 4 | 312 | |||
| BC038104.2 | - | 4 | 396 | 5269720 | ||

RCBTB1
Genomic Map