siRNA Details
| Gene Symbol: TMEM57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282564.1 | NP_001269493.1 | 7, 8 | 1188 | ||
| NM_018202.5 | NP_060672.2 | 9, 10 | 1869 | |||
| XM_005245931.2 | XP_005245988.1 | 10 | 1894 | |||
| XM_011541704.2 | XP_011540006.1 | 9, 10 | 1903 | |||
| GenBank mRNA | AK001609.1 | - | 9, 10 | 1780 | ||
| AY845015.1 | - | 9, 10 | 1799 | |||
| AY845035.1 | - | 7, 8 | 1056 | |||
| AY845036.1 | - | 4, 5 | 663 | |||
| BC032427.1 | - | 9, 10 | 1737 | 5246967 | ||

TMEM57
Genomic Map