siRNA Details
| Gene Symbol: TMEM57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282564.1 | NP_001269493.1 | 7 | 1073 | ||
| NM_018202.5 | NP_060672.2 | 9 | 1754 | |||
| XM_005245931.2 | XP_005245988.1 | 9 | 1715 | |||
| XM_011541704.2 | XP_011540006.1 | 9 | 1788 | |||
| GenBank mRNA | AK001609.1 | - | 9 | 1665 | ||
| AY845015.1 | - | 9 | 1684 | |||
| AY845035.1 | - | 7 | 941 | |||
| AY845036.1 | - | 4 | 548 | |||
| BC032427.1 | - | 9 | 1622 | 5246967 | ||

TMEM57
Genomic Map