siRNA Details
| Gene Symbol: SMU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018225.2 | NP_060695.2 | 10 | 1300 | ||
| XM_005251503.4 | XP_005251560.1 | 9 | 1136 | |||
| GenBank mRNA | AK001667.1 | - | 10 | 1230 | ||
| AK022032.1 | - | 2 | 104 | |||
| AK225559.1 | - | 10 | 1255 | |||
| AK225587.1 | - | 10 | 1249 | |||
| AK304767.1 | - | 8 | 887 | |||
| BC002876.2 | - | 10 | 1246 | 3940669 | ||
| BC110654.1 | - | 10 | 1340 | 6168993 | ||
| EF011613.1 | - | 10 | 1246 | |||

SMU1
Genomic Map