siRNA Details
| Gene Symbol: KIRREL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286349.1 | NP_001273278.1 | 5 | 934 | ||
| NM_018240.6 | NP_060710.3 | 7 | 1234 | |||
| XM_005245305.4 | XP_005245362.1 | 7 | 891 | |||
| GenBank mRNA | AK022708.1 | - | 3 | 236 | ||
| AK297788.1 | - | 5 | 562 | |||
| AY017369.1 | - | 7 | 830 | |||
| AY302131.1 | - | 5 | 929 | |||
| BC025268.1 | - | 2 | 82 | |||
| BC109192.1 | - | 7 | 897 | 40001501 | ||
| BC109193.1 | - | 7 | 897 | 40001502 | ||

KIRREL
Genomic Map