siRNA Details
| Gene Symbol: SLC47A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018242.2 | NP_060712.2 | 8 | 750 | ||
| GenBank mRNA | AK001709.1 | - | 8 | 750 | ||
| AK222625.1 | - | 8 | 754 | |||
| AK293196.1 | - | 1 | 43 | |||
| AK301854.1 | - | 7 | 707 | |||
| AK302620.1 | - | 7 | 652 | |||
| AK304647.1 | - | 1 | 43 | |||
| BC010661.2 | - | 8 | 739 | 3874616 | ||
| BC050592.1 | - | 8 | 724 | |||
| BC058882.1 | - | 8 | 726 | 5188651 | ||

SLC47A1
Genomic Map