siRNA Details
| Gene Symbol: ELP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242875.2 | NP_001229804.1 | 9 | 1007 | ||
| NM_001242876.2 | NP_001229805.1 | 8 | 929 | |||
| NM_001242877.2 | NP_001229806.1 | 7 | 734 | |||
| NM_001242878.2 | NP_001229807.1 | 7 | 734 | |||
| NM_001242879.2 | NP_001229808.1 | 5 | 602 | |||
| NM_001324465.1 | NP_001311394.1 | 8 | 812 | |||
| NM_001324466.1 | NP_001311395.1 | 8 | 929 | |||
| NM_001324467.1 | NP_001311396.1 | 8 | 812 | |||
| NM_001324468.1 | NP_001311397.1 | 7 | 733 | |||
| NM_018255.3 | NP_060725.1 | 8 | 812 | |||
| NR_040110.2 | 8 | 812 | ||||
| NR_136897.1 | 7 | 745 | ||||
| NR_136898.1 | 8 | 812 | ||||
| NR_137173.1 | 8 | 812 | ||||
| GenBank mRNA | AF332505.1 | - | 7 | 738 | ||
| AF403223.1 | - | 8 | 789 | |||
| AK001741.1 | - | 8 | 762 | |||
| AK022709.1 | - | 7 | 738 | |||
| AK222791.1 | - | 8 | 765 | |||
| AK293051.1 | - | 5 | 568 | |||
| AK300201.1 | - | 8 | 883 | |||
| AK302062.1 | - | 9 | 973 | |||
| BC032553.1 | - | 5 | 567 | 5555577 | ||
| CR457269.1 | - | 8 | 752 | |||

ELP2
Genomic Map