siRNA Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018266.2 | NP_060736.1 | 4, 5 | 839 | ||
| XM_005247578.1 | XP_005247635.1 | 4, 5 | 804 | |||
| XM_006713687.1 | XP_006713750.1 | 4, 5 | 747 | |||
| XM_011512958.2 | XP_011511260.1 | 4, 5 | 804 | |||
| GenBank mRNA | AK001764.1 | - | 4, 5 | 781 | ||
| AK222950.1 | - | 4, 5 | 771 | |||
| AK223248.1 | - | 4, 5 | 771 | |||
| BC021277.2 | - | 4, 5 | 757 | 4857987 | ||

TMEM39A
Genomic Map