siRNA Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018266.2 | NP_060736.1 | 8, 9 | 1665 | ||
| NR_073506.1 | 9, 10 | 1774 | ||||
| XM_005247578.1 | XP_005247635.1 | 7, 8 | 1509 | |||
| XM_006713687.1 | XP_006713750.1 | 8, 9 | 1573 | |||
| XM_017006788.1 | XP_016862277.1 | 7, 8 | 1546 | |||
| GenBank mRNA | AK001764.1 | - | 8, 9 | 1607 | ||
| AK222950.1 | - | 8, 9 | 1597 | |||
| AK223248.1 | - | 8, 9 | 1597 | |||
| BC003192.1 | - | 9, 10 | 1689 | |||
| BC010070.2 | - | 9, 10 | 1689 | |||
| BC021277.2 | - | 8, 9 | 1583 | 4857987 | ||

TMEM39A
Genomic Map