siRNA Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244676.1 | NP_001231605.1 | 6 | 1088 | ||
| NM_001244678.1 | NP_001231607.1 | 4 | 658 | |||
| NM_018271.4 | NP_060741.3 | 5 | 926 | |||
| XM_005264400.4 | XP_005264457.1 | 6 | 1038 | |||
| XM_005264401.4 | XP_005264458.1 | 6 | 1007 | |||
| XM_005264402.3 | XP_005264459.1 | 5 | 2827 | |||
| XM_005264403.4 | XP_005264460.1 | 5 | 2827 | |||
| XM_005264405.4 | XP_005264462.1 | 5 | 2827 | |||
| XM_006712041.3 | XP_006712104.1 | 5 | 2827 | |||
| XM_006712042.3 | XP_006712105.1 | 5 | 2827 | |||
| XM_006712043.2 | XP_006712106.1 | 5 | 2827 | |||
| XM_011532953.2 | XP_011531255.1 | 5 | 2827 | |||
| XM_011532954.2 | XP_011531256.1 | 4 | 2632 | |||
| XM_011532955.2 | XP_011531257.1 | 5 | 772 | |||
| GenBank mRNA | AK001778.1 | - | 5 | 546 | ||
| AK095303.1 | - | 5 | 2606 | |||
| AK289486.1 | - | 5 | 758 | |||
| AK315831.1 | - | 4 | 584 | |||
| AL832746.1 | - | 3 | 458 | |||
| BC035315.1 | - | 5 | 757 | |||
| BC047758.2 | - | 3 | 497 | |||
| BC064423.1 | - | 6 | 985 | 5752279 | ||
| HM185274.1 | - | 3 | 440 | |||

THNSL2
Genomic Map