siRNA Details
| Gene Symbol: NUDT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304745.1 | NP_001291674.1 | 2 | 446 | ||
| NM_018283.3 | NP_060753.1 | 2 | 446 | |||
| NR_136687.1 | 2 | 446 | ||||
| NR_136688.1 | 2 | 446 | ||||
| GenBank mRNA | AK001818.1 | - | 2 | 446 | ||
| BC050698.1 | - | 2 | 274 | |||
| BC064607.1 | - | 2 | 265 | |||
| BC107875.1 | - | 2 | 319 | 6451308 | ||
| BC133015.1 | - | 2 | 387 | 40147037 | ||
| BC133017.1 | - | 2 | 387 | 40147039 | ||
| DA000395.1 | - | 2 | 231 | |||
| DA760263.1 | - | 2 | 252 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172527, AK040749, BC060962, AK043196, AK032657, and others.

NUDT15
Genomic Map