siRNA Details
| Gene Symbol: NUDT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304745.1 | NP_001291674.1 | 2 | 421 | ||
| NM_018283.3 | NP_060753.1 | 2 | 421 | |||
| NR_136687.1 | 2 | 421 | ||||
| NR_136688.1 | 2 | 421 | ||||
| GenBank mRNA | AK001818.1 | - | 2 | 421 | ||
| BC050698.1 | - | 2 | 249 | |||
| BC064607.1 | - | 2 | 240 | |||
| BC107875.1 | - | 2 | 294 | 6451308 | ||
| BC133015.1 | - | 2 | 362 | 40147037 | ||
| BC133017.1 | - | 2 | 362 | 40147039 | ||
| DA000395.1 | - | 2 | 206 | |||
| DA760263.1 | - | 2 | 227 | |||

NUDT15
Genomic Map