siRNA Details
| Gene Symbol: TMEM140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018295.4 | NP_060765.4 | 2 | 423 | ||
| GenBank mRNA | AK001862.1 | - | 2 | 365 | ||
| AK225055.1 | - | 2 | 353 | |||
| AK301268.1 | - | 2 | 353 | |||
| BC020942.1 | - | 2 | 350 | 4798940 | ||
| DB064371.1 | - | 2 | 424 | |||
| DC323016.1 | - | 2 | 423 | |||

TMEM140
Genomic Map