siRNA Details
| Gene Symbol: KLHL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018316.1 | NP_060786.1 | 3 | 2289 | ||
| GenBank mRNA | AK001940.1 | - | 3 | 2289 | ||
| AK222656.1 | - | 4 | 2209 | |||
| AK294074.1 | - | 4 | 850 | |||
| BC026319.1 | - | 3 | 2283 | 4799495 | ||

KLHL26
Genomic Map