siRNA Details
| Gene Symbol: KLHL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018316.1 | NP_060786.1 | 3 | 2815 | ||
| GenBank mRNA | AK001940.1 | - | 3 | 2815 | ||
| BC026319.1 | - | 3 | 2809 | 4799495 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL26| Gene Symbol: KLHL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018316.1 | NP_060786.1 | 3 | 2815 | ||
| GenBank mRNA | AK001940.1 | - | 3 | 2815 | ||
| BC026319.1 | - | 3 | 2809 | 4799495 | ||