siRNA Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018342.4 | NP_060812.2 | 9 | 1169 | ||
| XM_005263110.1 | XP_005263167.1 | 10 | 1124 | |||
| XM_005263112.3 | XP_005263169.1 | 11 | 1159 | |||
| XM_006714254.1 | XP_006714317.1 | 10 | 981 | |||
| XM_017008366.1 | XP_016863855.1 | 8 | 1137 | |||
| XM_017008367.1 | XP_016863856.1 | 7 | 709 | |||
| XM_017008368.1 | XP_016863857.1 | 8 | 869 | |||
| GenBank mRNA | AK002017.1 | - | 8 | 1073 | ||
| BC026318.1 | - | 8 | 795 | |||
| BC054487.1 | - | 9 | 917 | 5732836 | ||
| BC060875.1 | - | 9 | 895 | 4800219 | ||

TMEM144
Genomic Map