siRNA Details
| Gene Symbol: ACER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300953.1 | NP_001287882.1 | 7 | 524 | ||
| NM_001300954.1 | NP_001287883.1 | 8 | 706 | |||
| NM_001300955.1 | NP_001287884.1 | 7 | 582 | |||
| NM_018367.6 | NP_060837.3 | 8 | 635 | |||
| XM_011545152.2 | XP_011543454.1 | 8 | 571 | |||
| GenBank mRNA | AF214454.1 | - | 8 | 589 | ||
| AF327353.1 | - | 8 | 600 | |||
| AK002100.1 | - | 10 | 766 | |||
| AK293800.1 | - | 9 | 606 | |||
| AK294978.1 | - | 7 | 455 | |||
| AK295142.1 | - | 8 | 623 | |||
| AK295327.1 | - | 9 | 719 | |||
| AK308856.1 | - | 10 | 763 | |||
| AK315000.1 | - | 8 | 601 | |||
| AK316120.1 | - | 7 | 520 | |||
| BC063034.1 | - | 9 | 595 | |||
| BC073853.1 | - | 8 | 552 | 3867010 | ||

ACER3
Genomic Map