siRNA Details
| Gene Symbol: DRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018370.2 | NP_060840.2 | 5, 6 | 1050 | ||
| XM_017019578.1 | XP_016875067.1 | 6, 7 | 1172 | |||
| GenBank mRNA | AK002121.1 | - | 3, 4 | 356 | ||
| AK094923.1 | - | 5, 6 | 680 | |||
| BC013773.1 | - | 3, 4 | 348 | |||
| BC018435.1 | - | 5, 6 | 1026 | 4474297 | ||

DRAM1
Genomic Map