siRNA Details
| Gene Symbol: NIPSNAP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018376.3 | NP_060846.2 | 2 | 660 | ||
| NR_130760.1 | 2 | 660 | ||||
| XM_011518839.1 | XP_011517141.1 | 2 | 660 | |||
| XM_017014889.1 | XP_016870378.1 | 2 | 612 | |||
| GenBank mRNA | AK002137.1 | - | 2 | 265 | ||
| BC005202.1 | - | 2 | 193 | 3933738 | ||
| BC017914.1 | - | 2 | 231 | 4292375 | ||
| BX647652.1 | - | 2 | 232 | |||

NIPSNAP3B
Genomic Map