siRNA Details
| Gene Symbol: PLCXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018390.3 | NP_060860.1 | 3 | 466 | ||
| NR_028057.1 | 4 | 449 | ||||
| XM_006724865.2 | XP_006724928.1 | 4 | 407 | |||
| XM_006724866.2 | XP_006724929.1 | 3 | 218 | |||
| XM_006724867.2 | XP_006724930.1 | 3 | 239 | |||
| XM_011545633.2 | XP_011543935.1 | 4 | 441 | |||
| GenBank mRNA | AK002185.1 | - | 4 | 451 | ||
| BC000888.1 | - | 3 | 372 | |||
| BC005028.1 | - | 3 | 398 | 3953383 | ||
| BC032072.1 | - | 3 | 372 | |||
| BU594555.1 | - | 3 | 466 | |||
| DA052662.1 | - | 3 | 463 | |||
| DA840761.1 | - | 3 | 246 | |||

PLCXD1
Genomic Map