siRNA Details
| Gene Symbol: PLCXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018390.3 | NP_060860.1 | 5 | 820 | ||
| NR_028057.1 | 6 | 803 | ||||
| XM_006724865.2 | XP_006724928.1 | 6 | 761 | |||
| XM_006724866.2 | XP_006724929.1 | 5 | 572 | |||
| XM_006724867.2 | XP_006724930.1 | 5 | 593 | |||
| XM_011545633.2 | XP_011543935.1 | 6 | 795 | |||
| GenBank mRNA | AK002185.1 | - | 6 | 805 | ||
| BC000888.1 | - | 5 | 726 | |||
| BC005028.1 | - | 5 | 752 | 3953383 | ||
| BC032072.1 | - | 5 | 726 | |||

PLCXD1
Genomic Map