siRNA Details
| Gene Symbol: SLC22A15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018420.2 | NP_060890.2 | 11 | 1660 | ||
| GenBank mRNA | AK125072.1 | - | 9 | 1228 | ||
| AL353933.1 | - | 3 | 361 | |||
| AY145501.1 | - | 11 | 1530 | |||
| BC026358.1 | - | 11 | 1659 | 4814675 | ||
| KU178698.1 | - | 11 | 1494 | |||
| KU178699.1 | - | 11 | 1752 | |||
| KU178700.1 | - | 10 | 1421 | |||
| KU178701.1 | - | 11 | 1418 | |||
| KU178702.1 | - | 11 | 1417 | |||
| KU178703.1 | - | 10 | 1281 | |||

SLC22A15
Genomic Map