siRNA Details
| Gene Symbol: HEMGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018437.4 | NP_060907.2 | 3 | 306 | ||
| NM_197978.2 | NP_932095.1 | 2 | 360 | |||
| XM_005252086.1 | XP_005252143.1 | 3 | 268 | |||
| XM_011518845.2 | XP_011517147.1 | 3 | 180 | |||
| XM_011518846.2 | XP_011517148.1 | 3 | 290 | |||
| GenBank mRNA | AF130060.2 | - | 2 | 93 | ||
| AF228713.2 | - | 3 | 213 | |||
| AF306682.1 | - | 2 | 142 | |||
| AF306683.1 | - | 2 | 108 | |||
| AF322875.1 | - | 3 | 212 | |||
| AK292441.1 | - | 3 | 232 | |||
| AL833094.1 | - | 2 | 153 | |||
| AY244805.1 | - | 2 | 306 | |||
| AY255672.1 | - | 2 | 211 | |||
| BC048324.1 | - | 3 | 218 | 5166612 | ||
| BG571634.1 | - | 2 | 184 | |||
| BG717036.1 | - | 3 | 248 | |||
| DA485094.1 | - | 2 | 360 | |||
| HY026328.1 | - | 3 | 307 | |||

HEMGN
Genomic Map