siRNA Details
| Gene Symbol: PIDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145886.3 | NP_665893.2 | 13 | 2232 | ||
| NM_145887.3 | NP_665894.2 | 13 | 2232 | |||
| XM_005253005.4 | XP_005253062.1 | 13 | 3242 | |||
| XM_005253006.4 | XP_005253063.1 | 13 | 2805 | |||
| XM_005253007.3 | XP_005253064.1 | 12 | 2322 | |||
| XM_005253008.4 | XP_005253065.1 | 13 | 3302 | |||
| XM_011520209.2 | XP_011518511.1 | 13 | 2468 | |||
| XM_011520210.2 | XP_011518512.1 | 14 | 3170 | |||
| XM_011520211.2 | XP_011518513.1 | 14 | 3170 | |||
| XM_011520212.1 | XP_011518514.1 | 12 | 2382 | |||
| XM_011520213.2 | XP_011518515.1 | 10 | 1524 | |||
| XM_017017993.1 | XP_016873482.1 | 10 | 1664 | |||
| GenBank mRNA | AB208832.1 | - | 10 | 3108 | ||
| AB208949.1 | - | 9 | 3413 | |||
| AB209529.1 | - | 11 | 3142 | |||
| AF229178.1 | - | 12 | 2401 | |||
| AF274972.1 | - | 13 | 2228 | |||
| AF465246.1 | - | 10 | 1612 | |||
| AK074893.1 | - | 10 | 1218 | |||
| AL833849.1 | - | 9 | 1540 | |||
| BC014904.1 | - | 12 | 2170 | 3908102 | ||

PIDD1
Genomic Map