siRNA Details
| Gene Symbol: NCBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114118.2 | NP_001107590.1 | 13 | 1851 | ||
| XM_017024826.1 | XP_016880315.1 | 12 | 1738 | |||
| XM_017024829.1 | XP_016880318.1 | 8 | 1143 | |||
| GenBank mRNA | AJ277841.1 | - | 8 | 1148 | ||
| AK125048.1 | - | 10 | 2540 | |||
| AK223470.1 | - | 6 | 1027 | |||
| BC010707.1 | - | 6 | 966 | |||

NCBP3
Genomic Map