siRNA Details
| Gene Symbol: SIRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039508.1 | NP_001034597.1 | 2 | 199 | ||
| NM_018556.3 | NP_061026.2 | 2 | 199 | |||
| NM_080816.2 | NP_543006.2 | 2 | 199 | |||
| XM_005260749.3 | XP_005260806.1 | 2 | 235 | |||
| XM_011529286.2 | XP_011527588.1 | 2 | 246 | |||
| XM_011529287.2 | XP_011527589.1 | 2 | 202 | |||
| GenBank mRNA | AB042624.1 | - | 2 | 215 | ||
| AK292745.1 | - | 2 | 199 | |||
| AY748247.1 | - | 2 | 134 | |||
| AY748248.1 | - | 2 | 134 | |||
| BC020629.1 | - | 2 | 176 | |||
| BC064532.1 | - | 2 | 160 | 5588038 | ||

SIRPG
Genomic Map