siRNA Details
| Gene Symbol: CHST12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243794.1 | NP_001230723.1 | 2 | 944 | ||
| NM_001243795.1 | NP_001230724.1 | 2 | 958 | |||
| NM_018641.4 | NP_061111.1 | 2 | 972 | |||
| XM_011515443.2 | XP_011513745.1 | 2 | 1097 | |||
| XM_011515444.2 | XP_011513746.1 | 2 | 920 | |||
| GenBank mRNA | AF239822.1 | - | 2 | 883 | ||
| AJ289131.1 | - | 1 | 870 | |||
| AK223191.1 | - | 2 | 893 | |||
| AK223211.1 | - | 2 | 893 | |||
| AK313484.1 | - | 2 | 893 | |||
| AY358574.1 | - | 2 | 908 | |||
| BC002918.1 | - | 2 | 925 | 3955396 | ||
| BC015954.1 | - | 2 | 925 | |||
| BC095492.1 | - | 2 | 932 | 30511863 | ||

CHST12
Genomic Map