siRNA Details
| Gene Symbol: SAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018666.2 | NP_061136.2 | 19 | 2726 | ||
| XM_017029621.1 | XP_016885110.1 | 19 | 2734 | |||
| XM_017029622.1 | XP_016885111.1 | 19 | 2645 | |||
| XM_017029623.1 | XP_016885112.1 | 19 | 2608 | |||
| XM_017029624.1 | XP_016885113.1 | 18 | 2548 | |||
| XM_017029625.1 | XP_016885114.1 | 18 | 2500 | |||
| XM_017029626.1 | XP_016885115.1 | 13 | 1850 | |||
| XM_017029627.1 | XP_016885116.1 | 9 | 1641 | |||
| GenBank mRNA | AJ278111.1 | - | 19 | 2726 | ||
| AK310412.1 | - | 17 | 3049 | |||
| BC144260.1 | - | 12 | 1479 | 9052780 | ||

SAGE1
Genomic Map