siRNA Details
| Gene Symbol: FEM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018708.2 | NP_061178.1 | 1 | 3087 | ||
| GenBank mRNA | AJ311359.1 | - | 1 | 2948 | ||
| AK074232.1 | - | 1 | 3043 | |||
| AK126532.1 | - | 2 | 2813 | |||
| AL359589.1 | - | 1 | 1227 | |||
| AL365408.1 | - | 1 | 1541 | |||
| AY356352.1 | - | 1 | 3041 | |||
| BC004988.1 | - | 1 | 3044 | 2957950 | ||
| CR749732.1 | - | 1 | 1227 | |||

FEM1A
Genomic Map