siRNA Details
| Gene Symbol: CKM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001824.4 | NP_001815.2 | 6, 7 | 957 | ||
| XM_017026240.1 | XP_016881729.1 | 5, 6 | 842 | |||
| GenBank mRNA | AK129878.1 | - | 6, 7 | 855 | ||
| AK300730.1 | - | 6, 7 | 745 | |||
| AK313281.1 | - | 6, 7 | 855 | |||
| BC007462.1 | - | 6, 7 | 855 | 3934232 | ||
| BT006793.1 | - | 5, 6 | 782 | |||
| M14780.1 | - | 6, 7 | 860 | |||
| M16440.1 | - | 14 | ||||

CKM
Genomic Map