siRNA Details
| Gene Symbol: FEM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018708.2 | NP_061178.1 | 1 | 1393 | ||
| GenBank mRNA | AJ311359.1 | - | 1 | 1253 | ||
| AK074232.1 | - | 1 | 1351 | |||
| AK126532.1 | - | 2 | 1121 | |||
| AK315554.1 | - | 1 | 1365 | |||
| AY356352.1 | - | 1 | 1347 | |||
| BC004988.1 | - | 1 | 1351 | 2957950 | ||

FEM1A
Genomic Map