siRNA Details
| Gene Symbol: ELMOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130037.1 | NP_001123509.1 | 6 | 740 | ||
| NM_001308018.1 | NP_001294947.1 | 7 | 876 | |||
| NM_018712.3 | NP_061182.3 | 6 | 740 | |||
| XM_017017994.1 | XP_016873483.1 | 7 | 871 | |||
| XM_017017995.1 | XP_016873484.1 | 6 | 621 | |||
| GenBank mRNA | AK054794.1 | - | 6 | 560 | ||
| AK297351.1 | - | 7 | 876 | |||
| AK303689.1 | - | 6 | 582 | |||
| BC028725.2 | - | 6 | 483 | 4815682 | ||

ELMOD1
Genomic Map