siRNA Details
| Gene Symbol: ELMOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130037.1 | NP_001123509.1 | 9, 10 | 1078 | ||
| NM_001308018.1 | NP_001294947.1 | 11, 12 | 1238 | |||
| NM_018712.3 | NP_061182.3 | 10, 11 | 1102 | |||
| XM_017017994.1 | XP_016873483.1 | 10, 11 | 1209 | |||
| XM_017017995.1 | XP_016873484.1 | 9, 10 | 959 | |||
| GenBank mRNA | AK054794.1 | - | 9, 10 | 898 | ||
| AK297351.1 | - | 11, 12 | 1238 | |||
| AK303689.1 | - | 9, 10 | 920 | |||
| AL359601.1 | - | 4, 5 | 289 | |||
| BC028725.2 | - | 10, 11 | 845 | 4815682 | ||

ELMOD1
Genomic Map