siRNA Details
| Gene Symbol: PLXNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017514.4 | NP_059984.3 | 2 | 329 | ||
| XM_005274705.4 | XP_005274762.2 | 2 | 323 | |||
| XM_006724829.3 | XP_006724892.1 | 2 | 323 | |||
| GenBank mRNA | CN427844.1 | - | 1 | 57 | ||
| HY098587.1 | - | 2 | 331 | |||
| HY158513.1 | - | 2 | 330 | |||
| X87852.1 | - | 2 | 338 | |||

PLXNA3
Genomic Map