siRNA Details
| Gene Symbol: GOLGA2P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036632.1 | 8, 9 | 1586 | |||
| GenBank mRNA | AF217995.1 | - | 8, 9 | 1536 | ||
| BC109389.1 | - | 4, 5 | 621 | |||
| BC110077.1 | - | 4, 5 | 621 | |||
| BC141930.1 | - | 4, 5 | 621 | 40029611 | ||
| BX647364.1 | - | 6 | 2320 | |||

GOLGA2P5
Genomic Map