siRNA Details
| Gene Symbol: ZCCHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017612.4 | NP_060082.2 | 3, 4 | 544 | ||
| XM_006719491.2 | XP_006719554.1 | 3, 4 | 460 | |||
| XM_011538555.2 | XP_011536857.1 | 2, 3 | 1086 | |||
| GenBank mRNA | AK027702.1 | - | 3, 4 | 413 | ||
| AK074638.1 | - | 3, 4 | 442 | |||
| AK307318.1 | - | 2, 3 | 277 | |||
| AL157433.1 | - | 2, 3 | 1838 | |||
| BC065918.1 | - | 3, 4 | 383 | 5752053 | ||
| DB456426.1 | - | 3, 4 | 467 | |||

ZCCHC8
Genomic Map