siRNA Details
| Gene Symbol: ZCCHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017612.4 | NP_060082.2 | 2, 3 | 485 | ||
| XM_006719491.2 | XP_006719554.1 | 2, 3 | 401 | |||
| GenBank mRNA | AK027702.1 | - | 2, 3 | 354 | ||
| AK074638.1 | - | 2, 3 | 383 | |||
| AL157433.1 | - | 1, 2 | 1779 | |||
| BC065918.1 | - | 2, 3 | 324 | 5752053 | ||
| DB048019.1 | - | 2, 3 | 486 | |||
| DB456426.1 | - | 2, 3 | 408 | |||

ZCCHC8
Genomic Map