siRNA Details
| Gene Symbol: DDX60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017631.5 | NP_060101.3 | 26 | 3807 | ||
| XM_011532103.2 | XP_011530405.1 | 26 | 3807 | |||
| GenBank mRNA | BC038115.2 | - | 26 | 3752 | 5261001 | |
| BX648655.1 | - | 6 | 648 | |||

DDX60
Genomic Map