siRNA Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286721.1 | NP_001273650.1 | 5 | 872 | ||
| NM_017664.3 | NP_060134.2 | 4 | 778 | |||
| NR_104586.1 | 5 | 910 | ||||
| NR_104587.1 | 5 | 872 | ||||
| GenBank mRNA | AK000100.1 | - | 4 | 740 | ||
| AK094222.1 | - | 1 | 778 | |||
| AK095763.1 | - | 2 | 305 | |||
| BC001727.1 | - | 5 | 771 | 3354202 | ||
| BC039715.1 | - | 5 | 840 | |||
| BX354283.2 | - | 5 | 769 | |||

ANKRD10
Genomic Map