siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 11 | 2078 | ||
| GenBank mRNA | AB105105.1 | - | 11 | 2078 | ||
| AF443278.1 | - | 11 | 1647 | |||
| AK000123.1 | - | 3 | 249 | |||
| AK092195.1 | - | 9 | 1136 | |||
| AK225216.1 | - | 3 | 249 | |||
| AK314394.1 | - | 11 | 1468 | |||
| AY137240.1 | - | 10 | 1289 | |||
| BC035882.1 | - | 11 | 1761 | 4941567 | ||
| BC040545.1 | - | 10 | 1333 | 5277060 | ||
| BX647465.1 | - | 6 | 485 | |||

FERMT1
Genomic Map