siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 14 | 2608 | ||
| GenBank mRNA | AB105105.1 | - | 14 | 2608 | ||
| AF443278.1 | - | 14 | 2177 | |||
| AK000123.1 | - | 6 | 779 | |||
| AK000747.1 | - | 4 | 474 | |||
| AK225216.1 | - | 6 | 779 | |||
| AK314394.1 | - | 14 | 1998 | |||
| AY137240.1 | - | 13 | 1819 | |||
| BC040545.1 | - | 13 | 1863 | 5277060 | ||
| BX647465.1 | - | 9 | 1016 | |||

FERMT1
Genomic Map