siRNA Details
| Gene Symbol: DEPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114120.2 | NP_001107592.1 | 2 | 421 | ||
| NM_017779.5 | NP_060249.2 | 2 | 421 | |||
| GenBank mRNA | AB281187.1 | - | 2 | 277 | ||
| AB281274.1 | - | 2 | 277 | |||
| AJ278112.1 | - | 2 | 277 | |||
| AK000361.1 | - | 2 | 282 | |||
| AK225735.1 | - | 2 | 267 | |||
| AK299324.1 | - | 2 | 282 | |||
| BC003511.1 | - | 2 | 270 | |||
| BC065304.1 | - | 2 | 224 | 6156084 | ||
| DB147703.1 | - | 2 | 421 | |||

DEPDC1
Genomic Map