siRNA Details
| Gene Symbol: DEPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114120.2 | NP_001107592.1 | 12 | 2607 | ||
| NM_017779.5 | NP_060249.2 | 11 | 1755 | |||
| GenBank mRNA | AB281187.1 | - | 12 | 2463 | ||
| AB281274.1 | - | 11 | 1611 | |||
| AJ278112.1 | - | 11 | 1611 | |||
| AK000490.1 | - | 5 | 932 | |||
| AK022792.1 | - | 4 | 1248 | |||
| AK225735.1 | - | 12 | 2453 | |||
| AL834208.1 | - | 13 | 1527 | |||
| BC065304.1 | - | 11 | 1558 | 6156084 | ||

DEPDC1
Genomic Map