siRNA Details
| Gene Symbol: BTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017797.3 | NP_060267.2 | 7 | 1221 | ||
| XM_005259593.1 | XP_005259650.1 | 7 | 986 | |||
| XM_011528127.1 | XP_011526429.1 | 5 | 587 | |||
| GenBank mRNA | AF355797.1 | - | 7 | 1221 | ||
| AK000393.1 | - | 4 | 1015 | |||
| AK027481.1 | - | 5 | 583 | |||
| AK056818.1 | - | 1 | 1356 | |||
| AK225831.1 | - | 7 | 1220 | |||
| AL360275.1 | - | 4 | 452 | |||
| AY007116.1 | - | 7 | 943 | |||
| BC000564.2 | - | 4 | 511 | |||
| BC008035.2 | - | 4 | 505 | |||
| BC053986.1 | - | 3 | 406 | |||
| BC069199.1 | - | 5 | 647 | |||
| BC094820.1 | - | 7 | 955 | |||
| CR457230.1 | - | 3 | 258 | |||

BTBD2
Genomic Map