siRNA Details
| Gene Symbol: BCAS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010974.2 | NP_001010974.1 | 2, 3 | 360 | ||
| NM_017843.4 | NP_060313.3 | 2, 3 | 360 | |||
| NM_198799.3 | NP_942094.2 | 2, 3 | 360 | |||
| XM_011528886.2 | XP_011527188.1 | 2, 3 | 283 | |||
| XM_011528887.2 | XP_011527189.1 | 2, 3 | 283 | |||
| XM_017027932.1 | XP_016883421.1 | 2, 3 | 283 | |||
| GenBank mRNA | AF361220.1 | - | 2, 3 | 360 | ||
| AJ511266.1 | - | 2, 3 | 184 | |||
| AK000502.1 | - | 2, 3 | 223 | |||
| BC038381.1 | - | 2, 3 | 210 | 4472674 | ||
| BC047337.1 | - | 2, 3 | 189 | |||

BCAS4
Genomic Map