siRNA Details
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 9 | 1445 | ||
NM_001193328.1 | NP_001180257.1 | 8 | 997 | |||
NM_017865.3 | NP_060335.2 | 9 | 1229 | |||
XM_011544220.1 | XP_011542522.1 | 9 | 1486 | |||
XM_011544221.1 | XP_011542523.1 | 9 | 1483 | |||
XM_011544222.1 | XP_011542524.1 | 9 | 1101 | |||
XM_011544223.2 | XP_011542525.1 | 8 | 1351 | |||
XM_011544227.1 | XP_011542529.1 | 9 | 1397 | |||
XM_017001698.1 | XP_016857187.1 | 9 | 1098 | |||
XM_017001699.1 | XP_016857188.1 | 8 | 1408 | |||
XM_017001700.1 | XP_016857189.1 | 8 | 1023 | |||
XM_017001701.1 | XP_016857190.1 | 9 | 1213 | |||
XM_017001702.1 | XP_016857191.1 | 8 | 1138 | |||
XM_017001703.1 | XP_016857192.1 | 9 | 1416 | |||
XM_017001704.1 | XP_016857193.1 | 9 | 1373 | |||
XM_017001705.1 | XP_016857194.1 | 9 | 1276 | |||
GenBank mRNA | AB209706.1 | - | 5 | 1283 | ||
AF256175.1 | - | 2 | 546 | |||
AK000538.1 | - | 8 | 966 | |||
AK001070.1 | - | 7 | 645 | |||
AK225535.1 | - | 8 | 966 | |||
AK298899.1 | - | 7 | 1174 | |||
AK302187.1 | - | 9 | 1379 | |||
AL133100.1 | - | 7 | 1958 | |||
BC002948.1 | - | 9 | 1185 | 3533691 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_182996, NM_001040686, AK170358, AK031177, BC150776, and others.
Panther Classification:
Gene Ontology Categories:
